The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284511.1 | NP_001271440.1 | 2 - 3 | 698 | 73 |
NM_001284513.1 | NP_001271442.1 | 2 - 3 | 321 | 73 | |
NM_001324118.1 | NP_001311047.1 | 2 - 3 | 815 | 73 | |
NM_152341.4 | NP_689554.2 | 2 - 3 | 815 | 73 | |
GenBank mRNA | AK054564.1 | - | 2 - 3 | 399 | 73 |
AK291373.1 | - | 2 - 3 | 517 | 73 | |
AY424282.1 | - | 2 - 3 | 385 | 73 | |
BC033703.1 | - | 2 - 3 | 473 | 73 | |
BQ935501.1 | - | 2 - 3 | 321 | 73 |