The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144061.1 | NP_001137533.1 | 10 - 11 | 1460 | 72 |
NM_001144062.1 | NP_001137534.1 | 10 - 11 | 1399 | 72 | |
NM_016451.4 | NP_057535.1 | 10 - 11 | 1521 | 72 | |
GenBank mRNA | AF084457.1 | - | 10 - 11 | 1359 | 72 |
AF111807.1 | - | 10 - 11 | 1327 | 72 | |
AK001203.1 | - | 4 - 5 | 468 | 72 | |
AL136593.1 | - | 10 - 11 | 1392 | 72 | |
BC037280.2 | - | 10 - 11 | 1515 | 72 |