The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144061.1 | NP_001137533.1 | 8 - 9 | 1202 | 127 |
NM_001144062.1 | NP_001137534.1 | 8 - 9 | 1141 | 127 | |
NM_016451.4 | NP_057535.1 | 8 - 9 | 1263 | 127 | |
GenBank mRNA | AF084457.1 | - | 8 - 9 | 1101 | 127 |
AF111807.1 | - | 8 - 9 | 1069 | 127 | |
AK001203.1 | - | 2 - 3 | 210 | 127 | |
AL136593.1 | - | 8 - 9 | 1134 | 127 | |
BC037280.2 | - | 8 - 9 | 1257 | 127 |