The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102401.2 | NP_001095871.1 | 2 - 3 | 842 | 69 |
NM_001265581.1 | NP_001252510.1 | 2 - 3 | 964 | 69 | |
NM_025115.3 | NP_079391.2 | 1 - 2 | 1211 | 69 | |
XM_011544665.2 | XP_011542967.1 | 2 - 3 | 1258 | 69 | |
GenBank mRNA | AK026916.1 | - | 1 - 2 | 1064 | 69 |
AL530920.3 | - | 1 - 2 | 740 | 69 | |
BC007387.2 | - | 2 - 3 | 767 | 69 | |
BC066935.1 | - | 2 - 3 | 795 | 69 | |
BX648410.1 | - | 2 - 3 | 878 | 69 |