The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102426.1 | NP_001095896.1 | 14 - 15 | 2534 | 60 |
XM_005263862.3 | XP_005263919.1 | 14 - 15 | 2823 | 60 | |
XM_005263863.4 | XP_005263920.1 | 14 - 15 | 2610 | 60 | |
XM_005263864.3 | XP_005263921.1 | 14 - 15 | 2775 | 60 | |
XM_011510507.1 | XP_011508809.1 | 16 - 17 | 2665 | 60 | |
XM_017003198.1 | XP_016858687.1 | 13 - 14 | 2730 | 60 | |
XM_017003199.1 | XP_016858688.1 | 13 - 14 | 2683 | 60 | |
GenBank mRNA | AB017114.1 | - | 10 - 11 | 1662 | 60 |
AB449889.1 | - | 14 - 15 | 2403 | 60 | |
AK292826.1 | - | 12 - 13 | 2167 | 60 |