The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308185.1 | NP_001295114.1 | 7 - 8 | 1546 | 152 |
NM_020307.3 | NP_064703.1 | 7 - 8 | 1546 | 152 | |
XM_005247648.3 | XP_005247705.1 | 8 - 9 | 1003 | 152 | |
XM_006713710.3 | XP_006713773.1 | 7 - 8 | 990 | 152 | |
XM_011513011.2 | XP_011511313.1 | 8 - 9 | 1555 | 152 | |
XM_017006891.1 | XP_016862380.1 | 8 - 9 | 1109 | 152 | |
XM_017006892.1 | XP_016862381.1 | 8 - 9 | 2675 | 152 | |
XM_017006893.1 | XP_016862382.1 | 8 - 9 | 2583 | 152 | |
XM_017006894.1 | XP_016862383.1 | 9 - 10 | 1140 | 152 | |
XM_017006895.1 | XP_016862384.1 | 9 - 10 | 1017 | 152 | |
XM_017006896.1 | XP_016862385.1 | 8 - 9 | 944 | 152 | |
XM_017006897.1 | XP_016862386.1 | 8 - 9 | 1078 | 152 | |
GenBank mRNA | AF180920.1 | - | 7 - 8 | 934 | 152 |
AF208843.1 | - | 3 - 4 | 367 | 152 | |
AF367476.1 | - | 9 - 10 | 1188 | 152 | |
AK022974.1 | - | 7 - 8 | 933 | 152 | |
AK122738.1 | - | 7 - 8 | 1546 | 152 | |
AY034790.1 | - | 8 - 9 | 1293 | 152 | |
AY358663.1 | - | 2 - 3 | 543 | 152 | |
BC007081.1 | - | 7 - 8 | 933 | 152 | |
BC038394.1 | - | 9 - 10 | 1196 | 152 | |
BC067812.1 | - | 7 - 8 | 922 | 152 | |
BX641146.1 | - | 2 - 3 | 3653 | 152 |