Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTR
Entrez Gene ID
4548
Gene Name
5-methyltetrahydrofolate-homocysteine methyltransferase
Gene Aliases
HMAG, MS, cblG
Chromosome Location
Chr.1: 236794304 - 236903981 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000254.2 NP_000245.2 3 - 4 766 70
NM_001291939.1 NP_001278868.1 3 - 4 766 70
NM_001291940.1 NP_001278869.1 3 - 4 766 70
XM_005273141.4 XP_005273198.1 3 - 4 735 70
XM_011544194.2 XP_011542496.1 2 - 3 1528 70
XM_017001329.1 XP_016856818.1 2 - 3 1528 70
XM_017001330.1 XP_016856819.1 2 - 3 1528 70
GenBank mRNA AK294270.1 - 2 - 3 520 70
AK299904.1 - 3 - 4 766 70
AK307390.1 - 3 - 4 755 70
BC130616.1 - 3 - 4 365 70
BC136440.1 - 3 - 4 365 70
BC144095.1 - 3 - 4 365 70
BC144097.1 - 3 - 4 365 70
BC144098.1 - 3 - 4 365 70
U71285.1 - 3 - 4 406 70
U73338.1 - 3 - 4 737 70
U75743.1 - 3 - 4 629 70

View Full Product Details