Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTR
Entrez Gene ID
4548
Gene Name
5-methyltetrahydrofolate-homocysteine methyltransferase
Gene Aliases
HMAG, MS, cblG
Chromosome Location
Chr.1: 236794304 - 236903981 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000254.2 NP_000245.2 7 - 8 1085 101
NM_001291939.1 NP_001278868.1 7 - 8 1085 101
NM_001291940.1 NP_001278869.1 7 - 8 1085 101
XM_005273141.4 XP_005273198.1 7 - 8 1054 101
XM_006711770.2 XP_006711833.1 3 - 4 196 101
XM_011544194.2 XP_011542496.1 6 - 7 1847 101
XM_017001329.1 XP_016856818.1 6 - 7 1847 101
XM_017001330.1 XP_016856819.1 6 - 7 1847 101
GenBank mRNA AK294270.1 - 7 - 8 896 101
AK299904.1 - 7 - 8 1085 101
AK307390.1 - 7 - 8 1074 101
AY429553.1 - 3 - 4 196 101
BC130616.1 - 7 - 8 684 101
BC136440.1 - 7 - 8 684 101
BC144095.1 - 7 - 8 684 101
BC144097.1 - 7 - 8 684 101
BC144098.1 - 7 - 8 684 101
U71285.1 - 7 - 8 725 101
U73338.1 - 7 - 8 1056 101
U75743.1 - 7 - 8 948 101

View Full Product Details