The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020361.4 | NP_065094.3 | 3 - 4 | 569 | 85 |
XM_017013647.1 | XP_016869136.1 | 3 - 4 | 1298 | 85 | |
GenBank mRNA | AF221594.1 | - | 3 - 4 | 526 | 85 |
BC033684.1 | - | 3 - 4 | 529 | 85 | |
BI756197.1 | - | 3 - 4 | 569 | 85 |