The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126132.2 | NP_001119604.1 | 6 - 7 | 333 | 87 |
NM_003283.5 | NP_003274.3 | 6 - 7 | 333 | 87 | |
XM_017027186.1 | XP_016882675.1 | 7 - 8 | 406 | 87 | |
GenBank mRNA | AK291352.1 | - | 6 - 7 | 185 | 87 |
AY762903.1 | - | 5 - 6 | 128 | 87 | |
BC010963.1 | - | 6 - 7 | 186 | 87 | |
BC034143.1 | - | 6 - 7 | 185 | 87 | |
DC385627.1 | - | 6 - 7 | 333 | 87 | |
DC385695.1 | - | 6 - 7 | 218 | 87 | |
M19309.1 | - | 6 - 7 | 185 | 87 | |
S69208.1 | - | 6 - 7 | 169 | 87 |