The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002944.2 | NP_002935.2 | 36 - 37 | 6145 | 91 |
XM_006715548.3 | XP_006715611.1 | 37 - 38 | 6237 | 91 | |
XM_011536049.2 | XP_011534351.1 | 37 - 38 | 6283 | 91 | |
XM_011536050.2 | XP_011534352.1 | 37 - 38 | 6280 | 91 | |
XM_011536051.2 | XP_011534353.1 | 36 - 37 | 6256 | 91 | |
XM_011536052.2 | XP_011534354.1 | 37 - 38 | 6241 | 91 | |
XM_011536053.2 | XP_011534355.1 | 37 - 38 | 6109 | 91 | |
XM_011536054.2 | XP_011534356.1 | 37 - 38 | 6284 | 91 | |
XM_017011172.1 | XP_016866661.1 | 36 - 37 | 6214 | 91 | |
XM_017011173.1 | XP_016866662.1 | 36 - 37 | 6313 | 91 | |
GenBank mRNA | BC144556.1 | - | 34 - 35 | 5327 | 91 |
M13880.1 | - | 3 - 4 | 389 | 91 | |
M34353.1 | - | 36 - 37 | 6152 | 91 |