Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LMTK3
Entrez Gene ID
114783
Gene Name
lemur tyrosine kinase 3
Gene Aliases
LMR3, PPP1R101, TYKLM3
Chromosome Location
Chr.19: 48485271 - 48513926 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080434.1 NP_001073903.1 7 - 8 736 63
XM_011526411.2 XP_011524713.1 7 - 8 1067 63
XM_011526412.2 XP_011524714.1 7 - 8 1034 63
GenBank mRNA AB067470.1 - 7 - 8 709 63

View Full Product Details