The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032583.3 | NP_115972.2 | 17 - 18 | 2381 | 58 |
NM_033151.3 | NP_149163.2 | 16 - 17 | 2667 | 58 | |
NM_145186.2 | NP_660187.1 | 17 - 18 | 2381 | 58 | |
XM_011523397.1 | XP_011521699.1 | 13 - 14 | 1887 | 58 | |
XM_011523398.2 | XP_011521700.1 | 5 - 6 | 602 | 58 | |
XM_017023795.1 | XP_016879284.1 | 17 - 18 | 4995 | 58 | |
XM_017023796.1 | XP_016879285.1 | 18 - 19 | 2468 | 58 | |
XM_017023797.1 | XP_016879286.1 | 18 - 19 | 2596 | 58 | |
XM_017023798.1 | XP_016879287.1 | 17 - 18 | 2422 | 58 | |
XM_017023799.1 | XP_016879288.1 | 17 - 18 | 2404 | 58 | |
XM_017023800.1 | XP_016879289.1 | 17 - 18 | 2395 | 58 | |
XM_017023801.1 | XP_016879290.1 | 16 - 17 | 4887 | 58 | |
XM_017023802.1 | XP_016879291.1 | 11 - 12 | 1742 | 58 | |
XM_017023803.1 | XP_016879292.1 | 17 - 18 | 4995 | 58 | |
GenBank mRNA | AF352582.1 | - | 17 - 18 | 2416 | 58 |
AF367202.1 | - | 17 - 18 | 2395 | 58 | |
AF411579.1 | - | 17 - 18 | 2395 | 58 | |
AY040219.1 | - | 16 - 17 | 2667 | 58 |