Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABCC11
Entrez Gene ID
85320
Gene Name
ATP binding cassette subfamily C member 11
Gene Aliases
EWWD, MRP8, WW
Chromosome Location
Chr.16: 48164842 - 48249973 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032583.3 NP_115972.2 18 - 19 2466 106
NM_033151.3 NP_149163.2 17 - 18 2752 106
NM_145186.2 NP_660187.1 18 - 19 2466 106
XM_011523397.1 XP_011521699.1 14 - 15 1972 106
XM_011523398.2 XP_011521700.1 6 - 7 687 106
XM_017023795.1 XP_016879284.1 18 - 19 5080 106
XM_017023796.1 XP_016879285.1 19 - 20 2553 106
XM_017023797.1 XP_016879286.1 19 - 20 2681 106
XM_017023798.1 XP_016879287.1 18 - 19 2507 106
XM_017023799.1 XP_016879288.1 18 - 19 2489 106
XM_017023800.1 XP_016879289.1 18 - 19 2480 106
XM_017023801.1 XP_016879290.1 17 - 18 4972 106
XM_017023802.1 XP_016879291.1 12 - 13 1827 106
XM_017023803.1 XP_016879292.1 18 - 19 5080 106
GenBank mRNA AF352582.1 - 18 - 19 2501 106
AF367202.1 - 18 - 19 2480 106
AF411579.1 - 18 - 19 2480 106
AK301547.1 - 17 - 18 2410 106
AY040219.1 - 17 - 18 2752 106

View Full Product Details