The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032583.3 | NP_115972.2 | 25 - 26 | 3605 | 66 |
NM_033151.3 | NP_149163.2 | 24 - 25 | 3891 | 66 | |
NM_145186.2 | NP_660187.1 | 25 - 26 | 3605 | 66 | |
XM_011523397.1 | XP_011521699.1 | 21 - 22 | 3111 | 66 | |
XM_011523398.2 | XP_011521700.1 | 13 - 14 | 1826 | 66 | |
XM_017023795.1 | XP_016879284.1 | 25 - 26 | 6219 | 66 | |
XM_017023796.1 | XP_016879285.1 | 26 - 27 | 3692 | 66 | |
XM_017023797.1 | XP_016879286.1 | 26 - 27 | 3820 | 66 | |
XM_017023798.1 | XP_016879287.1 | 25 - 26 | 3646 | 66 | |
XM_017023799.1 | XP_016879288.1 | 25 - 26 | 3628 | 66 | |
XM_017023800.1 | XP_016879289.1 | 25 - 26 | 3619 | 66 | |
XM_017023801.1 | XP_016879290.1 | 24 - 25 | 6111 | 66 | |
XM_017023802.1 | XP_016879291.1 | 19 - 20 | 2966 | 66 | |
GenBank mRNA | AF352582.1 | - | 25 - 26 | 3640 | 66 |
AF367202.1 | - | 25 - 26 | 3619 | 66 | |
AF411579.1 | - | 25 - 26 | 3619 | 66 | |
AY040219.1 | - | 24 - 25 | 3891 | 66 |