Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSEN34
Entrez Gene ID
79042
Gene Name
tRNA splicing endonuclease subunit 34
Gene Aliases
LENG5, PCH2C, SEN34, SEN34L
Chromosome Location
Chr.19: 54189517 - 54194536 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282332.1 NP_001269261.1 1 - 2 308 67
GenBank mRNA BC004530.2 - 1 - 2 308 67

View Full Product Details