Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSEN34
Entrez Gene ID
79042
Gene Name
tRNA splicing endonuclease subunit 34
Gene Aliases
LENG5, PCH2C, SEN34, SEN34L
Chromosome Location
Chr.19: 54189517 - 54194536 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011527294.2 XP_011525596.1 1 - 2 83 74

View Full Product Details