The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282333.1 | NP_001269262.1 | 1 - 2 | 107 | 57 |
XM_005278290.4 | XP_005278347.1 | 1 - 2 | 102 | 57 | |
GenBank mRNA | AK300054.1 | - | 1 - 2 | 88 | 57 |
DR763016.1 | - | 1 - 2 | 82 | 57 |