Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TACC2
Entrez Gene ID
10579
Gene Name
transforming acidic coiled-coil containing protein 2
Gene Aliases
AZU-1, ECTACC
Chromosome Location
Chr.10: 121989174 - 122254545 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291876.1 NP_001278805.1 16 - 17 8384 72
NM_001291877.1 NP_001278806.1 16 - 17 8531 72
NM_001291878.1 NP_001278807.1 13 - 14 3069 72
NM_001291879.1 NP_001278808.1 13 - 14 2742 72
NM_006997.3 NP_008928.1 13 - 14 3123 72
NM_206860.2 NP_996742.1 12 - 13 3033 72
NM_206861.2 NP_996743.1 16 - 17 3188 72
NM_206862.3 NP_996744.3 19 - 20 8750 72
XM_005269388.1 XP_005269445.1 20 - 21 8667 72
XM_005269389.4 XP_005269446.1 20 - 21 9191 72
XM_005269390.1 XP_005269447.1 21 - 22 8610 72
XM_005269391.1 XP_005269448.1 19 - 20 8607 72
XM_005269392.1 XP_005269449.1 19 - 20 8577 72
XM_005269393.1 XP_005269450.1 20 - 21 8574 72
XM_005269394.1 XP_005269451.1 19 - 20 8541 72
XM_005269395.1 XP_005269452.1 19 - 20 8532 72
XM_005269396.1 XP_005269453.1 18 - 19 8436 72
XM_005269397.1 XP_005269454.1 18 - 19 8406 72
XM_005269399.1 XP_005269456.1 17 - 18 8208 72
XM_006717548.1 XP_006717611.1 20 - 21 8562 72
XM_006717552.1 XP_006717615.1 19 - 20 8379 72
XM_006717553.1 XP_006717616.1 18 - 19 8343 72
XM_006717554.1 XP_006717617.1 18 - 19 8331 72
XM_011539126.1 XP_011537428.1 20 - 21 8664 72
XM_011539128.1 XP_011537430.1 18 - 19 3240 72
XM_011539129.1 XP_011537431.1 17 - 18 3105 72
XM_011539130.1 XP_011537432.1 17 - 18 3012 72
XM_011539131.1 XP_011537433.1 13 - 14 3078 72
XM_011539132.1 XP_011537434.1 14 - 15 3021 72
XM_011539133.1 XP_011537435.1 12 - 13 2695 72
XM_011539134.1 XP_011537436.1 12 - 13 2966 72
XM_017015484.1 XP_016870973.1 20 - 21 8655 72
XM_017015485.1 XP_016870974.1 20 - 21 8520 72
XM_017015486.1 XP_016870975.1 19 - 20 8484 72
XM_017015487.1 XP_016870976.1 19 - 20 8472 72
XM_017015488.1 XP_016870977.1 18 - 19 8301 72
XM_017015489.1 XP_016870978.1 17 - 18 8223 72
XM_017015491.1 XP_016870980.1 13 - 14 3218 72
XM_017015492.1 XP_016870981.1 13 - 14 3125 72
XM_017015493.1 XP_016870982.1 12 - 13 2883 72
XM_017015494.1 XP_016870983.1 11 - 12 2894 72
XM_017015495.1 XP_016870984.1 11 - 12 2754 72
XM_017015496.1 XP_016870985.1 14 - 15 3019 72
XM_017015497.1 XP_016870986.1 14 - 15 3278 72
XM_017015498.1 XP_016870987.1 13 - 14 3143 72
XM_017015499.1 XP_016870988.1 15 - 16 3112 72
GenBank mRNA AF095791.2 - 13 - 14 2710 72
AF176646.1 - 12 - 13 2868 72
AF220152.2 - 9 - 10 2556 72
AF528098.1 - 16 - 17 3169 72
AF528099.1 - 19 - 20 8731 72
AK027066.1 - 9 - 10 1662 72
AK075097.1 - 9 - 10 1020 72
AL713712.1 - 9 - 10 1968 72
AL833304.1 - 18 - 19 7849 72
BC000999.2 - 2 - 3 117 72
BC039311.1 - 12 - 13 3033 72
BC141837.1 - 14 - 15 2721 72
BC142673.1 - 14 - 15 2721 72
BC144599.1 - 16 - 17 8499 72
BC144600.1 - 16 - 17 8352 72
BC150627.1 - 16 - 17 8352 72

View Full Product Details