Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TACC2
Entrez Gene ID
10579
Gene Name
transforming acidic coiled-coil containing protein 2
Gene Aliases
AZU-1, ECTACC
Chromosome Location
Chr.10: 121989174 - 122254545 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291876.1 NP_001278805.1 8 - 9 7500 72
NM_001291877.1 NP_001278806.1 8 - 9 7647 72
NM_001291879.1 NP_001278808.1 5 - 6 1858 72
NM_006997.3 NP_008928.1 3 - 4 2008 72
NM_206860.2 NP_996742.1 3 - 4 2008 72
NM_206861.2 NP_996743.1 6 - 7 2073 72
NM_206862.3 NP_996744.3 9 - 10 7635 72
XM_005269388.1 XP_005269445.1 10 - 11 7459 72
XM_005269389.4 XP_005269446.1 10 - 11 7983 72
XM_005269391.1 XP_005269448.1 9 - 10 7399 72
XM_005269392.1 XP_005269449.1 10 - 11 7459 72
XM_005269393.1 XP_005269450.1 10 - 11 7459 72
XM_005269394.1 XP_005269451.1 9 - 10 7333 72
XM_005269395.1 XP_005269452.1 9 - 10 7324 72
XM_005269396.1 XP_005269453.1 10 - 11 7459 72
XM_005269397.1 XP_005269454.1 8 - 9 7198 72
XM_005269399.1 XP_005269456.1 9 - 10 7324 72
XM_006717553.1 XP_006717616.1 10 - 11 7459 72
XM_011539126.1 XP_011537428.1 10 - 11 7456 72
XM_011539128.1 XP_011537430.1 8 - 9 2032 72
XM_011539129.1 XP_011537431.1 7 - 8 1897 72
XM_011539130.1 XP_011537432.1 7 - 8 1897 72
XM_011539131.1 XP_011537433.1 3 - 4 1870 72
XM_011539133.1 XP_011537435.1 4 - 5 1811 72
XM_011539134.1 XP_011537436.1 4 - 5 2082 72
XM_017015486.1 XP_016870975.1 10 - 11 7459 72
XM_017015489.1 XP_016870978.1 8 - 9 7198 72
XM_017015491.1 XP_016870980.1 3 - 4 2010 72
XM_017015492.1 XP_016870981.1 3 - 4 2010 72
XM_017015494.1 XP_016870983.1 3 - 4 2010 72
XM_017015495.1 XP_016870984.1 3 - 4 1870 72
XM_017015496.1 XP_016870985.1 4 - 5 1811 72
GenBank mRNA AF095791.2 - 3 - 4 1595 72
AF220152.2 - 1 - 2 1672 72
AF528098.1 - 6 - 7 2054 72
AF528099.1 - 9 - 10 7616 72
AK027066.1 - 1 - 2 779 72
AK075097.1 - 1 - 2 136 72
AK295631.1 - 3 - 4 1791 72
AL713712.1 - 1 - 2 1084 72
AL833304.1 - 8 - 9 6734 72
BC039311.1 - 3 - 4 2008 72
BC144599.1 - 8 - 9 7615 72
BC144600.1 - 8 - 9 7468 72
BC150627.1 - 8 - 9 7468 72

View Full Product Details