Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TACC2
- Entrez Gene ID
-
10579
- Gene Name
- transforming acidic coiled-coil containing protein 2
- Gene Aliases
-
AZU-1, ECTACC
- Chromosome Location
-
Chr.10: 121989174 - 122254545 on Build GRCh38
View Full Product Details