The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000501.3 | NP_000492.2 | 18 - 19 | 1492 | 104 |
NM_001081752.2 | NP_001075221.1 | 17 - 18 | 1462 | 104 | |
NM_001081753.2 | NP_001075222.1 | 18 - 19 | 1507 | 104 | |
NM_001081754.2 | NP_001075223.1 | 18 - 19 | 1507 | 104 | |
NM_001081755.2 | NP_001075224.1 | 18 - 19 | 1492 | 104 | |
NM_001278912.1 | NP_001265841.1 | 18 - 19 | 1492 | 104 | |
NM_001278913.1 | NP_001265842.1 | 15 - 16 | 1384 | 104 | |
NM_001278914.1 | NP_001265843.1 | 17 - 18 | 1477 | 104 | |
NM_001278915.1 | NP_001265844.1 | 18 - 19 | 1492 | 104 | |
NM_001278916.1 | NP_001265845.1 | 17 - 18 | 1450 | 104 | |
NM_001278917.1 | NP_001265846.1 | 17 - 18 | 1462 | 104 | |
NM_001278918.1 | NP_001265847.1 | 15 - 16 | 1360 | 104 | |
NM_001278939.1 | NP_001265868.1 | 18 - 19 | 1492 | 104 | |
XM_005250187.1 | XP_005250244.1 | 17 - 18 | 1133 | 104 | |
XM_005250188.1 | XP_005250245.1 | 17 - 18 | 1125 | 104 | |
XM_011515868.1 | XP_011514170.1 | 18 - 19 | 1193 | 104 | |
XM_011515869.1 | XP_011514171.1 | 17 - 18 | 1122 | 104 | |
XM_011515870.1 | XP_011514172.1 | 17 - 18 | 1488 | 104 | |
XM_011515871.1 | XP_011514173.1 | 17 - 18 | 1151 | 104 | |
XM_011515873.1 | XP_011514175.1 | 18 - 19 | 1193 | 104 | |
XM_011515874.1 | XP_011514176.1 | 16 - 17 | 1458 | 104 | |
XM_011515875.1 | XP_011514177.1 | 16 - 17 | 1114 | 104 | |
XM_011515876.1 | XP_011514178.1 | 18 - 19 | 1193 | 104 | |
XM_017011813.1 | XP_016867302.1 | 16 - 17 | 1036 | 104 | |
XM_017011814.1 | XP_016867303.1 | 17 - 18 | 1151 | 104 | |
GenBank mRNA | AB208942.1 | - | 17 - 18 | 1088 | 104 |
AK075494.1 | - | 6 - 7 | 548 | 104 | |
AK075554.1 | - | 15 - 16 | 1067 | 104 | |
AK092232.1 | - | 15 - 16 | 975 | 104 | |
AK094138.1 | - | 17 - 18 | 1015 | 104 | |
AK095990.1 | - | 18 - 19 | 1127 | 104 | |
AK122731.1 | - | 12 - 13 | 755 | 104 | |
AK125511.1 | - | 17 - 18 | 1137 | 104 | |
AK130894.1 | - | 10 - 11 | 668 | 104 | |
AK225659.1 | - | 18 - 19 | 1106 | 104 | |
AK304845.1 | - | 18 - 19 | 1492 | 104 | |
BC035570.1 | - | 18 - 19 | 1097 | 104 | |
BC065566.1 | - | 17 - 18 | 1081 | 104 | |
BX537939.1 | - | 18 - 19 | 1127 | 104 | |
BX538199.1 | - | 17 - 18 | 1094 | 104 | |
M24782.1 | - | 9 - 10 | 604 | 104 | |
M36860.1 | - | 18 - 19 | 1142 | 104 | |
X52896.1 | - | 18 - 19 | 1141 | 104 |