The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000501.3 | NP_000492.2 | 7 - 8 | 776 | 79 |
NM_001081752.2 | NP_001075221.1 | 6 - 7 | 746 | 79 | |
NM_001081755.2 | NP_001075224.1 | 7 - 8 | 776 | 79 | |
NM_001278912.1 | NP_001265841.1 | 7 - 8 | 776 | 79 | |
NM_001278913.1 | NP_001265842.1 | 6 - 7 | 740 | 79 | |
NM_001278915.1 | NP_001265844.1 | 7 - 8 | 776 | 79 | |
NM_001278916.1 | NP_001265845.1 | 7 - 8 | 776 | 79 | |
NM_001278917.1 | NP_001265846.1 | 6 - 7 | 746 | 79 | |
NM_001278918.1 | NP_001265847.1 | 6 - 7 | 746 | 79 | |
NM_001278939.1 | NP_001265868.1 | 7 - 8 | 776 | 79 | |
XM_005250187.1 | XP_005250244.1 | 6 - 7 | 417 | 79 | |
XM_005250188.1 | XP_005250245.1 | 7 - 8 | 451 | 79 | |
XM_011515875.1 | XP_011514177.1 | 5 - 6 | 398 | 79 | |
XM_017011813.1 | XP_016867302.1 | 6 - 7 | 362 | 79 | |
GenBank mRNA | AB208942.1 | - | 6 - 7 | 372 | 79 |
AK075554.1 | - | 6 - 7 | 423 | 79 | |
AK092232.1 | - | 6 - 7 | 361 | 79 | |
AK094138.1 | - | 6 - 7 | 299 | 79 | |
AK225659.1 | - | 7 - 8 | 390 | 79 | |
AK304845.1 | - | 7 - 8 | 776 | 79 | |
BC035570.1 | - | 7 - 8 | 381 | 79 | |
BC050379.1 | - | 7 - 8 | 394 | 79 | |
BC065566.1 | - | 7 - 8 | 407 | 79 | |
BX538199.1 | - | 6 - 7 | 378 | 79 | |
M36860.1 | - | 7 - 8 | 426 | 79 | |
X52896.1 | - | 7 - 8 | 425 | 79 |