Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SMC1A
Entrez Gene ID
8243
Gene Name
structural maintenance of chromosomes 1A
Gene Aliases
CDLS2, DXS423E, SB1.8, SMC1, SMC1L1, SMC1alpha, SMCB
Chromosome Location
Chr.X: 53374149 - 53422728 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281463.1 NP_001268392.1 15 - 16 2583 74
NM_006306.3 NP_006297.2 14 - 15 2437 74
GenBank mRNA AK291921.1 - 14 - 15 2396 74
BC064368.1 - 14 - 15 2368 74
BC112127.1 - 14 - 15 2377 74
BC171726.1 - 15 - 16 2523 74
BX640922.1 - 14 - 15 2480 74
D80000.2 - 14 - 15 2309 74
S78271.1 - 14 - 15 2342 74

View Full Product Details