The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281463.1 | NP_001268392.1 | 9 - 10 | 1610 | 71 |
NM_006306.3 | NP_006297.2 | 8 - 9 | 1464 | 71 | |
GenBank mRNA | AK291921.1 | - | 8 - 9 | 1423 | 71 |
AK301266.1 | - | 5 - 6 | 688 | 71 | |
BC064368.1 | - | 8 - 9 | 1395 | 71 | |
BC112127.1 | - | 8 - 9 | 1404 | 71 | |
BC171726.1 | - | 9 - 10 | 1550 | 71 | |
BX640922.1 | - | 8 - 9 | 1507 | 71 | |
D80000.2 | - | 8 - 9 | 1336 | 71 | |
S78271.1 | - | 8 - 9 | 1369 | 71 |