Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDC14A
Entrez Gene ID
8556
Gene Name
cell division cycle 14A
Gene Aliases
DFNB105, cdc14, hCDC14
Chromosome Location
Chr.1: 100345015 - 100520281 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319210.1 NP_001306139.1 12 - 13 1735 65
NM_001319211.1 NP_001306140.1 12 - 13 1367 65
NM_001319212.1 NP_001306141.1 11 - 12 1647 65
NM_003672.3 NP_003663.2 12 - 13 1735 65
NM_033312.2 NP_201569.1 12 - 13 1735 65
XM_005271294.3 XP_005271351.1 12 - 13 1275 65
XM_005271296.3 XP_005271353.1 12 - 13 1272 65
XM_011542340.2 XP_011540642.1 12 - 13 1274 65
XM_011542341.2 XP_011540643.1 11 - 12 1215 65
XM_011542345.2 XP_011540647.1 9 - 10 1126 65
XM_017002646.1 XP_016858135.1 12 - 13 1275 65
XM_017002647.1 XP_016858136.1 12 - 13 1816 65
GenBank mRNA AB209857.1 - 9 - 10 1164 65
AF000367.1 - 12 - 13 1602 65
AF064102.1 - 12 - 13 1710 65
AF122013.1 - 12 - 13 1247 65
AK302310.1 - 12 - 13 1356 65
AK310292.1 - 12 - 13 1386 65
BC038979.1 - 11 - 12 1221 65
DQ530256.1 - 12 - 13 1253 65

View Full Product Details