Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDC14A
Entrez Gene ID
8556
Gene Name
cell division cycle 14A
Gene Aliases
DFNB105, cdc14, hCDC14
Chromosome Location
Chr.1: 100345015 - 100520281 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319210.1 NP_001306139.1 14 - 15 1905 78
NM_001319211.1 NP_001306140.1 14 - 15 1537 78
NM_001319212.1 NP_001306141.1 13 - 14 1817 78
NM_003672.3 NP_003663.2 14 - 15 1905 78
NM_033312.2 NP_201569.1 14 - 15 1905 78
XM_005271294.3 XP_005271351.1 14 - 15 1445 78
XM_005271296.3 XP_005271353.1 14 - 15 1442 78
XM_011542340.2 XP_011540642.1 14 - 15 1444 78
XM_011542341.2 XP_011540643.1 13 - 14 1385 78
XM_011542345.2 XP_011540647.1 11 - 12 1296 78
GenBank mRNA AB209857.1 - 11 - 12 1334 78
AF000367.1 - 14 - 15 1772 78
AF064102.1 - 14 - 15 1880 78
AF122013.1 - 14 - 15 1417 78
AK302310.1 - 14 - 15 1526 78
BC038979.1 - 13 - 14 1391 78
DQ530256.1 - 14 - 15 1423 78

View Full Product Details