Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDC14A
Entrez Gene ID
8556
Gene Name
cell division cycle 14A
Gene Aliases
DFNB105, cdc14, hCDC14
Chromosome Location
Chr.1: 100345015 - 100520281 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319210.1 NP_001306139.1 1 - 2 532 137
NM_001319212.1 NP_001306141.1 1 - 2 532 137
NM_003672.3 NP_003663.2 1 - 2 532 137
NM_033312.2 NP_201569.1 1 - 2 532 137
NM_033313.2 NP_201570.1 1 - 2 532 137
XM_017002647.1 XP_016858136.1 1 - 2 613 137
GenBank mRNA AF000367.1 - 1 - 2 399 136
AF064102.1 - 1 - 2 507 137
AF064103.1 - 1 - 2 497 137
AK310657.1 - 1 - 2 490 137
BC038979.1 - 1 - 2 106 137
BC093916.1 - 1 - 2 139 137
BC093918.1 - 1 - 2 139 137
DA885382.1 - 1 - 2 361 137
DQ530256.1 - 1 - 2 50 137

View Full Product Details