Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDC14A
Entrez Gene ID
8556
Gene Name
cell division cycle 14A
Gene Aliases
DFNB105, cdc14, hCDC14
Chromosome Location
Chr.1: 100345015 - 100520281 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319210.1 NP_001306139.1 2 - 3 633 73
NM_001319211.1 NP_001306140.1 2 - 3 265 73
NM_001319212.1 NP_001306141.1 2 - 3 633 73
NM_003672.3 NP_003663.2 2 - 3 633 73
NM_033312.2 NP_201569.1 2 - 3 633 73
NM_033313.2 NP_201570.1 2 - 3 633 73
XM_005271294.3 XP_005271351.1 2 - 3 173 73
XM_005271296.3 XP_005271353.1 2 - 3 170 73
XM_011542340.2 XP_011540642.1 2 - 3 172 73
XM_011542346.2 XP_011540648.1 2 - 3 181 73
XM_017002646.1 XP_016858135.1 2 - 3 173 73
XM_017002647.1 XP_016858136.1 2 - 3 714 73
GenBank mRNA AF000367.1 - 2 - 3 500 73
AF064102.1 - 2 - 3 608 73
AF064103.1 - 2 - 3 598 73
AF122013.1 - 2 - 3 145 73
AK302310.1 - 2 - 3 254 73
AK310657.1 - 2 - 3 591 73
BC038979.1 - 2 - 3 207 73
BC093916.1 - 2 - 3 240 73
BC093918.1 - 2 - 3 240 73
DA885382.1 - 2 - 3 462 73
DB029799.1 - 2 - 3 278 73
DQ530256.1 - 2 - 3 151 73

View Full Product Details