Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDC80
Entrez Gene ID
10403
Gene Name
NDC80, kinetochore complex component
Gene Aliases
HEC, HEC1, HsHec1, KNTC2, TID3, hsNDC80
Chromosome Location
Chr.18: 2571511 - 2616635 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006101.2 NP_006092.1 14 - 15 1739 133
GenBank mRNA AF017790.1 - 14 - 15 1661 133
AK289396.1 - 14 - 15 1693 133
AK313184.1 - 14 - 15 1691 133
BC035617.1 - 14 - 15 1670 133

View Full Product Details