The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278728.1 | NP_001265657.1 | 2 - 3 | 532 | 57 |
NM_134440.2 | NP_604389.1 | 3 - 4 | 694 | 57 | |
XM_005260136.4 | XP_005260193.1 | 3 - 4 | 658 | 57 | |
XM_005260137.4 | XP_005260194.1 | 3 - 4 | 429 | 57 | |
XM_006722930.3 | XP_006722993.1 | 2 - 3 | 527 | 57 | |
GenBank mRNA | AF105428.1 | - | 2 - 3 | 334 | 57 |
BG740813.1 | - | 2 - 3 | 387 | 57 | |
CX871218.1 | - | 3 - 4 | 627 | 57 |