The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_203503.1 | NP_987099.1 | 2 - 3 | 526 | 66 |
GenBank mRNA | AF325460.1 | - | 1 - 2 | 42 | 66 |