The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003693.3 | NP_003684.2 | 10 - 11 | 1728 | 145 |
NM_145350.2 | NP_663325.1 | 10 - 11 | 1691 | 145 | |
NR_028075.2 | 10 - 11 | 1658 | 145 | ||
NR_102409.1 | 10 - 11 | 1765 | 145 | ||
XM_017025256.1 | XP_016880745.1 | 6 - 7 | 629 | 145 | |
GenBank mRNA | AB052947.1 | - | 10 - 11 | 1592 | 145 |
AB052948.1 | - | 10 - 11 | 1658 | 145 | |
AB052949.1 | - | 9 - 10 | 2067 | 145 | |
AB052950.1 | - | 8 - 9 | 1394 | 145 | |
AK291810.1 | - | 10 - 11 | 1671 | 145 | |
AK298737.1 | - | 7 - 8 | 810 | 145 | |
AK310174.1 | - | 10 - 11 | 1688 | 145 | |
BC039735.1 | - | 10 - 11 | 1696 | 145 | |
D63483.1 | - | 10 - 11 | 1638 | 145 | |
D86864.1 | - | 10 - 11 | 1662 | 145 |