The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003800.1 | NP_001003800.1 | 4 - 5 | 1122 | 61 |
NM_015250.3 | NP_056065.1 | 4 - 5 | 1122 | 61 | |
XM_017014551.1 | XP_016870040.1 | 4 - 5 | 1243 | 61 | |
GenBank mRNA | AB014599.1 | - | 4 - 5 | 1042 | 61 |
AY052562.1 | - | 4 - 5 | 1122 | 61 | |
BC004296.1 | - | 1 - 2 | 353 | 61 | |
BC073970.1 | - | 4 - 5 | 1137 | 61 |