Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LCP2
Entrez Gene ID
3937
Gene Name
lymphocyte cytosolic protein 2
Gene Aliases
SLP-76, SLP76
Chromosome Location
Chr.5: 170248084 - 170297818 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005565.3 NP_005556.1 1 - 2 279 70
GenBank mRNA AK300000.1 - 1 - 2 258 70
AK310369.1 - 1 - 2 220 70
BC016618.1 - 1 - 2 216 70
BG911397.1 - 1 - 2 279 70
BT007273.1 - 1 - 2 72 70
FN432830.1 - 1 - 2 72 70
U20158.1 - 1 - 2 327 70

View Full Product Details