Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCL11A
Entrez Gene ID
53335
Gene Name
B-cell CLL/lymphoma 11A
Gene Aliases
BCL11A-L, BCL11A-S, BCL11A-XL, BCL11a-M, CTIP1, EVI9, HBFQTL5, ZNF856
Chromosome Location
Chr.2: 60451167 - 60553498 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018014.3 NP_060484.2 2 - 3 615 122
NM_022893.3 NP_075044.2 2 - 3 615 122
NM_138559.1 NP_612569.1 2 - 3 615 122
XM_011532909.1 XP_011531211.1 2 - 3 615 122
XM_011532910.1 XP_011531212.1 2 - 3 615 122
XM_017004333.1 XP_016859822.1 2 - 3 383 122
GenBank mRNA AB058712.1 - 2 - 3 515 122
AK055251.1 - 2 - 3 615 122
BC021098.1 - 2 - 3 496 122

View Full Product Details