The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320613.1 | NP_001307542.1 | 1 - 2 | 334 | 52 |
NM_004036.4 | NP_004027.2 | 1 - 2 | 334 | 52 | |
XM_006711925.1 | XP_006711988.1 | 1 - 2 | 50 | 52 | |
XM_011532489.1 | XP_011530791.1 | 1 - 2 | 50 | 52 | |
XM_011532490.1 | XP_011530792.1 | 1 - 2 | 50 | 52 | |
XM_011532491.1 | XP_011530793.1 | 1 - 2 | 50 | 52 | |
XM_011532492.2 | XP_011530794.1 | 1 - 2 | 50 | 52 | |
XM_011532493.1 | XP_011530795.1 | 1 - 2 | 50 | 52 | |
XM_011532494.1 | XP_011530796.1 | 1 - 2 | 50 | 52 | |
XM_017003186.1 | XP_016858675.1 | 1 - 2 | 50 | 52 | |
XM_017003187.1 | XP_016858676.1 | 1 - 2 | 50 | 52 | |
XM_017003188.1 | XP_016858677.1 | 1 - 2 | 50 | 52 | |
XM_017003189.1 | XP_016858678.1 | 1 - 2 | 50 | 52 | |
XM_017003190.1 | XP_016858679.1 | 1 - 2 | 50 | 52 | |
GenBank mRNA | BM148122.1 | - | 1 - 2 | 156 | 52 |
HY136273.1 | - | 1 - 2 | 339 | 52 |