Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PUM2
Entrez Gene ID
23369
Gene Name
pumilio RNA binding family member 2
Gene Aliases
PUMH2, PUML2
Chromosome Location
Chr.2: 20248692 - 20352423 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282752.1 NP_001269681.1 9 - 10 1455 84
NM_001282790.1 NP_001269719.1 9 - 10 1318 84
NM_001282791.1 NP_001269720.1 10 - 11 1443 84
NM_015317.2 NP_056132.1 10 - 11 1564 84
XM_005262607.1 XP_005262664.1 10 - 11 1538 84
XM_005262609.1 XP_005262666.1 10 - 11 1538 84
XM_005262610.1 XP_005262667.1 10 - 11 1538 84
XM_006711973.1 XP_006712036.1 10 - 11 1335 84
XM_006711974.2 XP_006712037.1 10 - 11 1375 84
XM_006711975.2 XP_006712038.1 11 - 12 1499 84
XM_011532719.1 XP_011531021.1 12 - 13 1734 84
XM_011532720.2 XP_011531022.1 12 - 13 1743 84
XM_011532721.2 XP_011531023.1 11 - 12 1535 84
XM_011532723.1 XP_011531025.1 9 - 10 1165 84
XM_011532724.2 XP_011531026.1 10 - 11 1538 84
XM_011532725.2 XP_011531027.1 10 - 11 1538 84
XM_017003701.1 XP_016859190.1 10 - 11 1335 84
XM_017003702.1 XP_016859191.1 10 - 11 1375 84
XM_017003703.1 XP_016859192.1 11 - 12 1563 84
XM_017003704.1 XP_016859193.1 13 - 14 1881 84
XM_017003705.1 XP_016859194.1 13 - 14 1868 84
XM_017003706.1 XP_016859195.1 12 - 13 1660 84
XM_017003707.1 XP_016859196.1 10 - 11 1335 84
XM_017003708.1 XP_016859197.1 10 - 11 1568 84
XM_017003709.1 XP_016859198.1 11 - 12 1534 84
XM_017003710.1 XP_016859199.1 10 - 11 1384 84
XM_017003711.1 XP_016859200.1 10 - 11 1335 84
XM_017003712.1 XP_016859201.1 10 - 11 1403 84
XM_017003713.1 XP_016859202.1 11 - 12 1562 84
XM_017003714.1 XP_016859203.1 6 - 7 944 84
XM_017003715.1 XP_016859204.1 9 - 10 1165 84
GenBank mRNA AF272350.1 - 7 - 8 965 84
AF315591.1 - 9 - 10 1315 84
AK093847.1 - 10 - 11 1416 84
AK304198.1 - 9 - 10 1225 84
BC112046.1 - 9 - 10 1318 84
BC112048.1 - 9 - 10 1318 84
BC143550.1 - 9 - 10 1318 84
BC143552.1 - 10 - 11 1443 84
D87078.3 - 9 - 10 1292 84

View Full Product Details