Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PUM2
Entrez Gene ID
23369
Gene Name
pumilio RNA binding family member 2
Gene Aliases
PUMH2, PUML2
Chromosome Location
Chr.2: 20248692 - 20352423 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282752.1 NP_001269681.1 13 - 14 2383 109
NM_001282790.1 NP_001269719.1 12 - 13 2009 109
NM_001282791.1 NP_001269720.1 13 - 14 2134 109
NM_015317.2 NP_056132.1 14 - 15 2492 109
XM_005262607.1 XP_005262664.1 14 - 15 2466 109
XM_005262609.1 XP_005262666.1 13 - 14 2229 109
XM_005262610.1 XP_005262667.1 13 - 14 2229 109
XM_006711973.1 XP_006712036.1 14 - 15 2263 109
XM_006711974.2 XP_006712037.1 14 - 15 2303 109
XM_006711975.2 XP_006712038.1 15 - 16 2427 109
XM_011532719.1 XP_011531021.1 16 - 17 2662 109
XM_011532720.2 XP_011531022.1 16 - 17 2671 109
XM_011532721.2 XP_011531023.1 15 - 16 2463 109
XM_011532723.1 XP_011531025.1 13 - 14 2093 109
XM_017003701.1 XP_016859190.1 14 - 15 2263 109
XM_017003702.1 XP_016859191.1 14 - 15 2303 109
XM_017003703.1 XP_016859192.1 15 - 16 2491 109
XM_017003704.1 XP_016859193.1 17 - 18 2809 109
XM_017003705.1 XP_016859194.1 17 - 18 2796 109
XM_017003706.1 XP_016859195.1 16 - 17 2588 109
XM_017003707.1 XP_016859196.1 13 - 14 2026 109
XM_017003708.1 XP_016859197.1 13 - 14 2259 109
XM_017003709.1 XP_016859198.1 14 - 15 2225 109
XM_017003710.1 XP_016859199.1 13 - 14 2075 109
XM_017003711.1 XP_016859200.1 13 - 14 2026 109
XM_017003712.1 XP_016859201.1 13 - 14 2094 109
XM_017003713.1 XP_016859202.1 14 - 15 2253 109
XM_017003714.1 XP_016859203.1 10 - 11 1872 109
XM_017003715.1 XP_016859204.1 12 - 13 1856 109
GenBank mRNA AF272350.1 - 10 - 11 1656 109
AF315591.1 - 13 - 14 2243 109
AK093847.1 - 14 - 15 2344 109
AK304198.1 - 13 - 14 2153 109
BC024218.2 - 2 - 3 489 109
BC112046.1 - 13 - 14 2246 109
BC112048.1 - 13 - 14 2246 109
BC143550.1 - 12 - 13 2009 109
BC143552.1 - 13 - 14 2134 109
D87078.3 - 13 - 14 2220 109

View Full Product Details