Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PUM2
Entrez Gene ID
23369
Gene Name
pumilio RNA binding family member 2
Gene Aliases
PUMH2, PUML2
Chromosome Location
Chr.2: 20248692 - 20352423 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282752.1 NP_001269681.1 6 - 7 1045 87
NM_001282790.1 NP_001269719.1 6 - 7 908 87
NM_001282791.1 NP_001269720.1 7 - 8 1033 87
NM_015317.2 NP_056132.1 7 - 8 1154 87
XM_005262607.1 XP_005262664.1 7 - 8 1128 87
XM_005262609.1 XP_005262666.1 7 - 8 1128 87
XM_005262610.1 XP_005262667.1 7 - 8 1128 87
XM_006711973.1 XP_006712036.1 7 - 8 925 87
XM_006711974.2 XP_006712037.1 7 - 8 965 87
XM_006711975.2 XP_006712038.1 8 - 9 1089 87
XM_011532719.1 XP_011531021.1 9 - 10 1324 87
XM_011532720.2 XP_011531022.1 9 - 10 1333 87
XM_011532721.2 XP_011531023.1 8 - 9 1125 87
XM_011532723.1 XP_011531025.1 6 - 7 755 87
XM_011532724.2 XP_011531026.1 7 - 8 1128 87
XM_011532725.2 XP_011531027.1 7 - 8 1128 87
XM_017003701.1 XP_016859190.1 7 - 8 925 87
XM_017003702.1 XP_016859191.1 7 - 8 965 87
XM_017003703.1 XP_016859192.1 8 - 9 1153 87
XM_017003704.1 XP_016859193.1 10 - 11 1471 87
XM_017003705.1 XP_016859194.1 10 - 11 1458 87
XM_017003706.1 XP_016859195.1 9 - 10 1250 87
XM_017003707.1 XP_016859196.1 7 - 8 925 87
XM_017003708.1 XP_016859197.1 7 - 8 1158 87
XM_017003709.1 XP_016859198.1 8 - 9 1124 87
XM_017003710.1 XP_016859199.1 7 - 8 974 87
XM_017003711.1 XP_016859200.1 7 - 8 925 87
XM_017003712.1 XP_016859201.1 7 - 8 993 87
XM_017003713.1 XP_016859202.1 8 - 9 1152 87
XM_017003714.1 XP_016859203.1 3 - 4 534 87
XM_017003715.1 XP_016859204.1 6 - 7 755 87
GenBank mRNA AF272350.1 - 4 - 5 555 87
AF315591.1 - 6 - 7 905 87
AK093847.1 - 7 - 8 1006 87
AK304198.1 - 6 - 7 815 87
BC112046.1 - 6 - 7 908 87
BC112048.1 - 6 - 7 908 87
BC143550.1 - 6 - 7 908 87
BC143552.1 - 7 - 8 1033 87
D87078.3 - 6 - 7 882 87

View Full Product Details