The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138328.2 | NP_612201.1 | 1 - 2 | 128 | 92 |
XM_017024272.1 | XP_016879761.1 | 1 - 2 | 133 | 92 | |
GenBank mRNA | AJ313480.1 | - | 1 - 2 | 128 | 92 |
BC100975.1 | - | 1 - 2 | 127 | 92 | |
BC100976.1 | - | 1 - 2 | 128 | 92 |