The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130386.2 | NP_569057.1 | 2 - 3 | 273 | 148 |
GenBank mRNA | AB005145.1 | - | 2 - 3 | 128 | 148 |
AB034251.1 | - | 1 - 2 | 58 | 148 | |
AB038518.1 | - | 2 - 3 | 266 | 148 | |
BC060789.1 | - | 2 - 3 | 245 | 148 |