The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130386.2 | NP_569057.1 | 7 - 8 | 2163 | 102 |
XM_011525741.2 | XP_011524043.1 | 6 - 7 | 2021 | 102 | |
GenBank mRNA | AB005145.1 | - | 7 - 8 | 2018 | 102 |
AB034251.1 | - | 6 - 7 | 1948 | 102 | |
AB038518.1 | - | 7 - 8 | 2156 | 102 | |
AB052103.1 | - | 5 - 6 | 3546 | 102 | |
AL713657.1 | - | 3 - 4 | 975 | 102 | |
BC060789.1 | - | 7 - 8 | 2135 | 102 |