Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDC80
Entrez Gene ID
10403
Gene Name
NDC80, kinetochore complex component
Gene Aliases
HEC, HEC1, HsHec1, KNTC2, TID3, hsNDC80
Chromosome Location
Chr.18: 2571511 - 2616635 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006101.2 NP_006092.1 1 - 2 176 66
GenBank mRNA AF017790.1 - 1 - 2 98 66
AK289396.1 - 1 - 2 130 66
BC005239.1 - 1 - 2 146 66
BC010171.2 - 1 - 2 104 66
BC035617.1 - 1 - 2 107 66

View Full Product Details