The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098614.2 | NP_001092084.1 | 1 - 2 | 71 | 131 |
XM_017019997.1 | XP_016875486.1 | 1 - 2 | 84 | 131 | |
GenBank mRNA | CR627435.1 | - | 1 - 2 | 71 | 131 |