The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170553.1 | NP_001164024.1 | 5 - 6 | 840 | 102 |
NM_182607.4 | NP_872413.1 | 4 - 5 | 732 | 102 | |
XM_005262127.4 | XP_005262184.1 | 6 - 7 | 942 | 102 | |
XM_011530936.2 | XP_011529238.1 | 6 - 7 | 801 | 102 | |
GenBank mRNA | AK292472.1 | - | 4 - 5 | 611 | 102 |
AK301311.1 | - | 5 - 6 | 740 | 102 | |
AK301783.1 | - | 4 - 5 | 731 | 102 | |
AK315846.1 | - | 4 - 5 | 637 | 102 | |
BC043216.2 | - | 4 - 5 | 688 | 102 | |
BX640913.1 | - | 4 - 5 | 646 | 102 | |
BX648658.1 | - | 4 - 5 | 732 | 102 |