Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MGA
Entrez Gene ID
23269
Gene Name
MGA, MAX dimerization protein
Gene Aliases
MAD5, MXD5
Chromosome Location
Chr.15: 41621139 - 41769943 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080541.2 NP_001074010.2 9 - 10 3610 251
NM_001164273.1 NP_001157745.1 9 - 10 3610 251
XM_005254243.2 XP_005254300.1 9 - 10 3610 251
XM_005254244.3 XP_005254301.1 9 - 10 3537 251
XM_005254245.4 XP_005254302.1 9 - 10 3767 251
XM_005254246.2 XP_005254303.1 9 - 10 3656 251
XM_005254247.2 XP_005254304.1 9 - 10 3555 251
XM_005254248.2 XP_005254305.1 9 - 10 3609 251
XM_005254249.2 XP_005254306.1 9 - 10 3609 251
XM_005254252.2 XP_005254309.1 9 - 10 3609 251
XM_005254253.2 XP_005254310.1 9 - 10 3609 251
XM_006720443.3 XP_006720506.1 9 - 10 3574 251
XM_006720445.3 XP_006720508.1 8 - 9 2439 251
XM_011521397.2 XP_011519699.1 9 - 10 3609 251
XM_011521398.2 XP_011519700.1 9 - 10 3609 251
XM_011521401.2 XP_011519703.1 9 - 10 3609 251
XM_017022028.1 XP_016877517.1 8 - 9 2482 251
XM_017022029.1 XP_016877518.1 8 - 9 2425 251
XM_017022030.1 XP_016877519.1 9 - 10 3608 251
XM_017022031.1 XP_016877520.1 8 - 9 2482 251
XM_017022033.1 XP_016877522.1 9 - 10 3608 251
GenBank mRNA AK025751.1 - 7 - 8 1667 251
BC136659.1 - 8 - 9 3499 251

View Full Product Details