The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281463.1 | NP_001268392.1 | 4 - 5 | 682 | 70 |
NM_006306.3 | NP_006297.2 | 3 - 4 | 536 | 70 | |
GenBank mRNA | AK291921.1 | - | 3 - 4 | 495 | 70 |
BC064368.1 | - | 3 - 4 | 467 | 70 | |
BC080185.1 | - | 3 - 4 | 471 | 70 | |
BC112127.1 | - | 3 - 4 | 476 | 70 | |
BC171726.1 | - | 4 - 5 | 622 | 70 | |
BP312805.1 | - | 3 - 4 | 536 | 70 | |
BX640922.1 | - | 3 - 4 | 579 | 70 | |
D80000.2 | - | 3 - 4 | 408 | 70 | |
S78271.1 | - | 3 - 4 | 441 | 70 |