Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DST
Entrez Gene ID
667
Gene Name
dystonin
Gene Aliases
BP240, BPA, BPAG1, CATX-15, CATX15, D6S1101, DMH, DT, EBSB2, HSAN6, MACF2
Chromosome Location
Chr.6: 56457987 - 56954671 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144769.2 NP_001138241.1 24 - 25 3273 107
NM_001144770.1 NP_001138242.1 22 - 23 2944 107
NM_001723.5 NP_001714.1 11 - 12 1828 107
NM_015548.4 NP_056363.2 11 - 12 1828 107
NM_183380.3 NP_899236.1 21 - 22 3218 107
XM_005249310.3 XP_005249367.1 25 - 26 3367 107
XM_005249315.3 XP_005249372.1 24 - 25 3314 107
XM_005249316.2 XP_005249373.1 17 - 18 3590 107
XM_005249318.2 XP_005249375.1 23 - 24 3028 107
XM_005249319.2 XP_005249376.1 22 - 23 2929 107
XM_005249320.4 XP_005249377.1 22 - 23 2985 107
XM_005249322.4 XP_005249379.1 22 - 23 2880 107
XM_005249323.3 XP_005249380.1 21 - 22 3213 107
XM_005249324.2 XP_005249381.1 11 - 12 1825 107
XM_011514824.2 XP_011513126.1 24 - 25 3278 107
XM_011514825.2 XP_011513127.1 24 - 25 3274 107
XM_011514826.2 XP_011513128.1 21 - 22 3213 107
XM_017011205.1 XP_016866694.1 25 - 26 3377 107
XM_017011206.1 XP_016866695.1 25 - 26 3377 107
XM_017011207.1 XP_016866696.1 25 - 26 3314 107
XM_017011208.1 XP_016866697.1 25 - 26 3377 107
XM_017011209.1 XP_016866698.1 25 - 26 3377 107
XM_017011210.1 XP_016866699.1 25 - 26 3377 107
XM_017011211.1 XP_016866700.1 25 - 26 3377 107
XM_017011212.1 XP_016866701.1 25 - 26 3377 107
XM_017011213.1 XP_016866702.1 25 - 26 3377 107
XM_017011214.1 XP_016866703.1 25 - 26 3377 107
XM_017011215.1 XP_016866704.1 25 - 26 3377 107
XM_017011216.1 XP_016866705.1 25 - 26 3377 107
XM_017011217.1 XP_016866706.1 22 - 23 2958 107
XM_017011219.1 XP_016866708.1 25 - 26 3373 107
XM_017011220.1 XP_016866709.1 24 - 25 3316 107
XM_017011221.1 XP_016866710.1 25 - 26 3373 107
XM_017011222.1 XP_016866711.1 17 - 18 3532 107
XM_017011223.1 XP_016866712.1 25 - 26 3373 107
XM_017011224.1 XP_016866713.1 21 - 22 3213 107
XM_017011225.1 XP_016866714.1 25 - 26 3387 107
GenBank mRNA AK299898.1 - 22 - 23 2944 107
AU117550.1 - 1 - 2 91 107
BC065536.1 - 11 - 12 1792 107
L11690.1 - 11 - 12 1822 107

View Full Product Details