Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DST
Entrez Gene ID
667
Gene Name
dystonin
Gene Aliases
BP240, BPA, BPAG1, CATX-15, CATX15, D6S1101, DMH, DT, EBSB2, HSAN6, MACF2
Chromosome Location
Chr.6: 56457987 - 56954671 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144769.2 NP_001138241.1 55 - 56 10286 71
NM_001144770.1 NP_001138242.1 53 - 54 9957 71
NM_015548.4 NP_056363.2 42 - 43 8841 71
NM_183380.3 NP_899236.1 52 - 53 10231 71
XM_005249310.3 XP_005249367.1 61 - 62 16638 71
XM_005249315.3 XP_005249372.1 60 - 61 16585 71
XM_005249316.2 XP_005249373.1 53 - 54 16861 71
XM_005249318.2 XP_005249375.1 59 - 60 16299 71
XM_005249319.2 XP_005249376.1 58 - 59 16200 71
XM_005249320.4 XP_005249377.1 58 - 59 16256 71
XM_005249322.4 XP_005249379.1 58 - 59 16151 71
XM_005249323.3 XP_005249380.1 57 - 58 16484 71
XM_005249324.2 XP_005249381.1 47 - 48 15096 71
XM_011514824.2 XP_011513126.1 60 - 61 16549 71
XM_011514825.2 XP_011513127.1 55 - 56 10287 71
XM_011514826.2 XP_011513128.1 57 - 58 16484 71
XM_017011205.1 XP_016866694.1 61 - 62 16648 71
XM_017011206.1 XP_016866695.1 61 - 62 16648 71
XM_017011207.1 XP_016866696.1 61 - 62 16585 71
XM_017011208.1 XP_016866697.1 61 - 62 16648 71
XM_017011209.1 XP_016866698.1 61 - 62 16648 71
XM_017011210.1 XP_016866699.1 61 - 62 16648 71
XM_017011211.1 XP_016866700.1 61 - 62 16648 71
XM_017011212.1 XP_016866701.1 60 - 61 16414 71
XM_017011213.1 XP_016866702.1 61 - 62 16648 71
XM_017011214.1 XP_016866703.1 61 - 62 16648 71
XM_017011215.1 XP_016866704.1 61 - 62 16648 71
XM_017011216.1 XP_016866705.1 61 - 62 16648 71
XM_017011217.1 XP_016866706.1 58 - 59 16229 71
XM_017011218.1 XP_016866707.1 23 - 24 12257 71
XM_017011219.1 XP_016866708.1 56 - 57 10386 71
XM_017011220.1 XP_016866709.1 55 - 56 10329 71
XM_017011221.1 XP_016866710.1 55 - 56 10152 71
XM_017011222.1 XP_016866711.1 48 - 49 10545 71
XM_017011223.1 XP_016866712.1 56 - 57 10386 71
XM_017011224.1 XP_016866713.1 52 - 53 10226 71
GenBank mRNA AF400226.1 - 42 - 43 8733 71
AK055189.1 - 2 - 3 1571 71

View Full Product Details